Learn More
BD Vacutainer™ K2EDTA Tubes
Product Code. 15744718
Change view
Click to view available options
Capacity (Metric):
2 mL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15744718 2 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15744718 Supplier BD Supplier No. 368841
  • ONLINE EXCLUSIVE PRICE
    5715.00 NOK
    5450.94 NOK / Pack of 1000
    Save 264.06 NOK
    4% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Specifications
Product Type K2 EDTA Tube
Additive K2 EDTA
Material PET
Closure Type Hemogard
Capacity (Metric) 2 mL
Closure Color Pink
Tube Size 13 x 75 mm
For Use With (Application) Used to obtain either a serum sample or a whole blood or EDTA plasma sample
Label Type Paper
compliance-icons
Product Identifier
  • X1000 Vacutainer K2EDTA 2ML Purple
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: dipotassium EDTA dihydrate