Learn More
BD Vacutainer™ Hemogard™ Closure Plastic K2-Edta Tube
Product Code. 10331254
Change view
Click to view available options
Capacity (Metric):
10 mL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10331254 10 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10331254 Supplier BD Supplier No. 367525
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used to obtain a whole blood or EDTA plasma sample. Vacutainer™ Hemogard™ Closure Plastic K2-Edta Tube contains K2EDTA which is a liquid solution that is spray coated onto the interior surface of the BD Vacutainer Plus Tubes.
Lavender Hemogard™ closure
Paper label
TRUSTED_SUSTAINABILITY
Specifications
Product Type K2 EDTA Tube
Additive K2 EDTA
Material Plastic
Closure Type Hemogard
Capacity (Metric) 10 mL
Closure Color Lavender
Height (Metric) 100 mm
For Use With (Application) Used to obtain a whole blood or EDTA plasma sample
Label Type Paper
Diameter (Metric) Outer 16 mm
Certifications/Compliance ISO 6710
Show More Show Less
compliance-icons
Product Identifier
  • X1000 BD Vacutainer 16 x 100 mm x 10.0 mL
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: dipotassium EDTA dihydrate