Learn More
Greiner Bio-One VACUETTE™ FC Mix Blood Collection Tube
Product Code. 16387756
Change view
Click to view available options
Capacity (Metric):
3 mL
Closure Color:
Pink with Black Ring
Unit Size:
Pack of 1200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color Unit Size
16387756 3 mL Pink with Black Ring Pack of 1200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16387756 Supplier Greiner Bio-One Supplier No. 454513
  • ONLINE EXCLUSIVE PRICE
    5882.49 NOK
    4309.33 NOK / Pack of 1200
    Save 1573.16 NOK
    26% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Vacuette™ Blood Collection Tubes have set the standard in today‘s marketplace. They provide improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass.
  • Made from virtually unbreakable PET plastic
  • Optically clear as glass
  • Effective glycolysis inhibition for precise determination of blood sugar content
  • Powder additives ensure no dilution effects
  • Improved safety and hygiene during specimen collection procedures
TRUSTED_SUSTAINABILITY
Specifications
Additive Sodium Fluoride EDTA
Material PET
Sterility Sterile
Capacity (Metric) 3 mL
Closure Color Pink with Black Ring
Tube Size 13 mm x 75 mm
For Use With (Application) Blood glucose analysis
Label Type Transparent
compliance-icons compliance-icons
Product Identifier
  • Vacuette FC Mix Tube
Signal Word
  • Danger||Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • serious eye damage/eye irritation
  • Acute toxicity Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /