Learn More
MMM Medcenter™ Vacucell Vacuum Drying Oven
Description
- Temperature sensitive, easily decomposing or oxidable samples can be gently dried in vacuum drying oven, where air displacement is obtained by inert gas
- Complicated components with difficult to access cavities dry quickly
- Temperature range up to 250°C
- Microprocessor control
- Interface RS 232
- Three programmes
- Timer max. 99 h 59 min
Specifications
Specifications
| Capacity (Metric) | 111 L |
| Temperature (Metric) | 250°C |
| Type | Vacuum Drying Oven |
| Timer | 99 hr 59 min |
| Dimensions (D x W x H) Interior | 410 x 540 x 580 mm |
| Width (Metric) | 760 mm |
| Height (Metric) | 880 mm |
| Depth (Metric) | 640 mm |