Learn More
Cytiva VacuCap™ 60 Vacuum Filtration Devices
GREENER_CHOICE
Product Code. 17134821
Change view
Click to view available options
Pore Size:
0.2 μm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Unit Size
17134821 0.2 μm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17134821 Supplier Cytiva Supplier No. TA4632
  • ONLINE EXCLUSIVE PRICE
    4920.00 NOK
    4638.48 NOK / Pack of 10
    Save 281.52 NOK
    5% Off
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
Remove endotoxin from water, buffer, neutral sugars, and certain biological solutions.
  • Not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Connectors 1/4 in. hose barb inner taper
Material (Membrane) Hydrophilic PES
Pore Size 0.2 μm
Sterility Sterile
No. per Pack 10/Pk.
Diameter (Metric) 60 mm