Description
For sampling very small quantities of highly potent, expensive or active powders for analysis. The smallest sample quantities from 0.1 ml to 2.5 ml can be collected with the PharmaPicker. By simply exchanging the volume tip in the collection cylinder for 0.1–1.2 ml or 1.25–2.5 ml, the sampling volume is modified.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Capacity (Metric) | 0.1 to 1.2 mL |
| Product Type | PharmaPicker |
| Finish | Electro-polished |
| For Use With (Application) | Small Quantities of Powder |
| Includes | Collection Cylinder |
| Material | Stainless Steel (V4A) |
| Handle Type | T-shape |
| Length (Metric) | 60 cm |