Learn More
Brady™ V4500 Barcode Scanner Kit
Product Code. 30057756
Click to view available options
Includes:
V4500 Barcode Scanner
V4500 Barcode Scanner, V1200 Battery
V4500 Barcode Scanner, V1300 Cradle, V1200 Battery, AC Adapter, Power Cable, USB 1 m Cable
Unit Size:
1 set
Each
Product Code. 30057756
Brand: Brady™ 177229
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Rugged design. Seamless experience. Digitize data capture and automate your workflows with ease and accuracy with the V4500 Barcode Scanner.
  • High-speed, omnidirectional barcode scanning made simple.
  • Ideal for inventory management, kit verification, scan and print, and packout applications.
  • Proprietary Cortex Decoder technology allows scanner to easily read tiny barcodes as well as shiny and hard-to-read surfaces.
  • Automated keyboard device and dynamic user interface lets you go from setup to first scan in seconds.
  • Data parsing feature lets you sort and arrange data exactly how you want – start with a simple scan and easily connect to the cloud and your ERP system.
  • Withstands 6 ft. drops (MIL-STD-810G).
Specifications
Description V4500 Barcode Reader Kit
Battery Run Time Up to 75,000 scans per charge
Supported Barcode Types BC412, Codabar, Code 11, Code 128, Code 32, Code 39, Code 93, GS1 DataBar, Hong Kong 2 of 5, IATA 2 of 5, Interleaved 2 of 5, Matrix 2 of 5, MSI Plessey, NEC 2 of 5, Pharmacode, Plessey, Straight 2 of 5, Telepen, UPC / EAN / JAN, Trioptic
Data Transfer Method Bluetooth™ 5 Low Energy (class II)
Height (Metric) 75 mm
Height (English) 2.95 in.
Width (Metric) 152 mm
Width (English) 5.98 in.
Depth (Metric) 113 mm
Depth (English) 4.449 in.
Weight (Metric) 1.38 kg
Charging Time Charging over cradle (0 to 100 %) 4 hr.
Weight (English) 3.042 lb.
Certifications/Compliance CE, UKCA
For Use With (Application) Asset tracking, Barcode Labelling, Circuit Board Labelling, General Identification
Field View High Density Field: 30° Horizontal by 20° Vertical; Wide Field: 50° Horizontal by 33.50° Vertical
Humidity 5% to 95% (non-condensing)
Includes V4500 Barcode Scanner, V1300 Cradle, V1200 Battery, AC Adapter, Power Cable, USB 1 m Cable
Warranty 3 Years
Type Barcode Reader
Connection Type RS-232, USB
Resolution 1280 x 960
Show More Show Less
Has lithium-ion battery - must ship ground