Description
- Optimum integration and use of space – with a depth of 86 cm, V-LINE cabinets now use the space next to laboratory fume cupboards and work benches that conventional hazardous material cabinets have left unused
- Fire resistance of 90 minutes
- Maximum comfort – vertical drawer technology allows access and visibility to all stored containers from both sides
- At the push of a button the vertical drawers open and close automatically
- Drawers glide softly and quietly out of and into the cabinet body without additional physical efforts
- The stop function of the vertical drawers detects resistances and eliminates the risk of being trapped
- No unauthorized use – doors lockable with cylinder lock and cabinets can be integrated by the customer into an existing master key system
- Locking status indicator (red/green) provided
- Always closed – serial automatic door closing after approx. 60 seconds with acoustic/visual signal before closing
- Possible interior equipment includes height-adjustable shelves, bottom collecting sumps and guard rail systems
- In accordance with EN 14470-1 and TRGS 510
- Robust construction and longevity – scratch-resistant outer body with powder-coated surface
- Ventilation – integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Specifications
| Certifications/Compliance | EN 14470-1, TRGS 510 |
| Configuration | 4 x 50 kg sheet steel shelves; 1 x 50 kg sheet steel insert; 1 x 13 L sheet steel sump |
| Dimensions (D x W x H) Interior | 775 x 331 x 1750 mm |
| Dimensions (D x W x H) | 860 x 449 x 1966 mm |
| Door Style | Auto-close |
| For Use With (Application) | Storage of flammable hazardous substances |
| Load Bearing Capacity | 4 x Shelves: 50 kg each, Insert: 50 kg, Sump: 13 L, Maximum load: 120 kg |
| No. of Doors | 1 |
| Weight (Metric) | 394.5 kg |
| Color | Anthracite Gray, Warning Yellow |
| Show More |