Learn More
Asecos™ V-MOVE-90 Safety Storage Cabinets
Product Code. 15826501 Shop All asecos Products
Change view
Click to view available options
Load Bearing Capacity:
4 x Shelves: 50 kg each, Insert: 50 kg, Sump: 13 L, Maximum load: 120 kg
4 x Shelves: 50 kg each, Inserts: 50 kg each, Sumps: 13 L each, Maximum load: 240 kg
5 x Shelves: 50 kg each, Inserts: 50 kg each, Sumps: 13 L each, Maximum load: 240 kg
Unit Size:
Each
Product Code. Load Bearing Capacity unitSize
15826501 4 x Shelves: 50 kg each, Inserts: 50 kg each, Sumps: 13 L each, Maximum load: 240 kg Each
15939541 5 x Shelves: 50 kg each, Inserts: 50 kg each, Sumps: 13 L each, Maximum load: 240 kg Each
15916371 4 x Shelves: 50 kg each, Insert: 50 kg, Sump: 13 L, Maximum load: 120 kg Each
3 options
Product Code. 15826501
Brand: Asecos™ 3508604135104
This item requires a price update prior to being ordered.
Request a Price for this item here.
For the safe and approved storage of flammable hazardous substances in working areas with electronic open/close doors.
  • Optimum integration and use of space – with a depth of 86 cm, V-LINE cabinets now use the space next to laboratory fume cupboards and work benches that conventional hazardous material cabinets have left unused
  • Fire resistance of 90 minutes
  • Maximum comfort – vertical drawer technology allows access and visibility to all stored containers from both sides
  • At the push of a button the vertical drawers open and close automatically
  • Drawers glide softly and quietly out of and into the cabinet body without additional physical efforts
  • The stop function of the vertical drawers detects resistances and eliminates the risk of being trapped
  • No unauthorized use – doors lockable with cylinder lock and cabinets can be integrated by the customer into an existing master key system
  • Locking status indicator (red/green) provided
  • Always closed – serial automatic door closing after approx. 60 seconds with acoustic/visual signal before closing
  • Possible interior equipment includes height-adjustable shelves, bottom collecting sumps and guard rail systems
  • In accordance with EN 14470-1 and TRGS 510
  • Robust construction and longevity – scratch-resistant outer body with powder-coated surface
  • Ventilation – integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Certifications/Compliance EN 14470-1, TRGS 510
Configuration 4 x 50 kg sheet steel shelves; 1 x 50 kg sheet steel insert; 1 x 13 L sheet steel sump; 5 x 50 kg sheet steel shelves; 1 x 50 kg sheet steel insert; 1 x 13 L sheet steel sump
Dimensions (D x W x H) Interior 775 x 701 x 1750 mm
Dimensions (D x W x H) 860 x 819 x 1966 mm
Door Style Auto-close
For Use With (Application) Storage of flammable hazardous substances
Load Bearing Capacity 4 x Shelves: 50 kg each, Inserts: 50 kg each, Sumps: 13 L each, Maximum load: 240 kg
No. of Doors 2
Weight (Metric) 604 kg
Color Anthracite Gray, Light Gray
Material Gypsum
Product Type Flammables Storage Cabinet