Learn More
UVP UV Light Converter Plates
Description
- Convert UV to 460-470nm with the Visi-Blue™ plate for viewing GFP stains
- Scratch-resistant glass is mounted in a metal housing for durability
- Rubber seal stops the plate from sliding
- Handles are situated on two sides of the plate for easy handling
- Several standard plate sizes are available
- Variety of wavelengths for gel documentation, gel excitation and gel imaging
Specifications
Specifications
| Product Type | Converter Plate |
| For Use With (Application) | Convert 302nm UV to 460 to 470nm Blue Light |
| Filtration Area (English) | 8.26 x 10.23 in. |
| Filtration Area (Metric) | 21 x 26 cm |
| Model | Visi-Blue Converter Plates |