Learn More
UVP C-15G Mini Chromato-Vue™ UV Viewing Cabinet
Description
- Designed with a metal base and plastic top
- Includes a UVGL-58 UV lamp
- The bottom panel of the cabinet is removable
- The lamp fits snugly on top of the cabinet
- A contrast control filter absorbs UV energy and protects the eyes
- Soft rubber viewport provides a comfortable viewing area with space for eyeglasses
- Lightweight access curtain blocks out external light from the cabinet interior
- Lamps for other mini cabinets are ordered separately
Specifications
Specifications
| Voltage | 230V |
| Depth (Metric) | 229mm |
| Height (Metric) | 305mm |
| Width (Metric) | 267mm |
| Model | C-15G |
| Packaging Quantity | 1Pack |
| For Use With (Application) | Mineralogy, Lab Fluorescence, Sterilization, Field Investigation |
| Product Type | UV Cabinet |