Learn More
UVP Replacement UV Tubes
Artikelnummer. 11798231
Ansicht ändern
Click to view available options
Wavelength:
254 nm
302 nm
365 nm
Packungsgröße:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Wavelength Packungsgröße
11798231 302 nm Each
12862989 365 nm Each
12822989 254 nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Artikelnummer. 11798231 Lieferant UVP Lieferanten-Nr. 34004201
Estimated Shipment: 01-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
For use with 8w UV lamps.
For use with 8w UV lamps.
  • For use with 8w UV lamps
Spezifikation
Wattage 8 W
Wavelength 302 nm
For Use With (Equipment) CL-1000M crosslinker, TMW-20 and benchtop series transilluminator and EL series UV lamp
Product Type Replacement UV Tube