Learn More
UVP Replacement UV Light Tubes
Product Code. 12813858
Change view
Click to view available options
Wavelength:
302 nm
365 nm
Wattage:
6 W
15 W
20 W
40 W
Unit Size:
Each
Product Code. Wavelength Wattage unitSize
12813858 365 nm 40 W Each
12842989 302 nm 6 W Each
12893848 365 nm 20 W Each
12883918 365 nm 15 W Each
4 options
Product Code. 12813858
Brand: UVP 34006802
Only null left
Add to basket
Add to basket
Replacement UV light tubes for 4, 6, 15, 20 and 40 watt ultraviolet lamps and transilluminators
Specifications
Wattage 40 W
Wavelength 365 nm
For Use With (Equipment) Model XX-40 UV bench lamp
Packaging Quantity 1 Ea.