Learn More
UVP Portable UV Lamp Replacement Tubes
Product Code. 11747993
Click to view available options
Wavelength:
365 nm
365/284 nm
Unit Size:
Each
Product Code. 11747993
Brand: UVP 34001501
Only null left
Add to basket
Add to basket
Provide a variety of uses including laboratory, research, or criminology applications. UVP™ Portable UV Lamp Replacement Tubes are for use with Model ML-49 Portable UV Lamp with Handle.
Lamps are lightweight for easy handling
  • Durable plastic housing
  • Six watts
  • Models available with shortwave, longwave UV or a combination shortwave/longwave
  • Combination shortwave/longwave version comes with a snap-on wavelength selector for switching between wavelengths
Specifications
Light Source UV-A
Wattage 6 W
Wavelength 365/284 nm
For Use With (Equipment) Tube multiband shortwave / longwave accessory to handheld UV lamps 6W
Packaging Quantity 1 Ea.
Product Type UV Lamp