Learn More
UVP MRL-58 Multiple Ray Lamp Tube
Description
- UV tube
- 8w bulb
- 365nm
Applications:
Gel electrophoresis, Nucleic acid visualization, DNA analysis, Biochemical testing
Specifications
Specifications
| Wattage | 8 W |
| Wavelength | 365 nm |
| For Use With (Equipment) | MRL-58 multiple ray lamp and Visi-Blue™ transilluminator |
| Length (English) | 11.33 in. |
| Length (Metric) | 287.7 mm |
| Packaging Quantity | 1 Ea. |