Learn More
UVP Compact and Handheld UV Lamp Replacement Tubes
Product Code. p-168209
Click to view available options
Wavelength:
254 nm
365 nm
Wattage:
4 W
6 W
Unit Size:
Each
Product Code. 12812979
Brand: UVP 34000301
Only null left
Add to basket
Add to basket
For use with 4w and 6w UV lamps.
  • For use with 4w and 6w UV lamps
Specifications
Light Source UV-C
Voltage 115 V
Wattage 4 W
Wavelength 254 nm
For Use With (Equipment) EL-series UV lamp, 3UV™ lamp, UVG-4 mini UV lamp, UVG-11, UVGL-25 compact UV lamp, DE-1 and DE-4 Memorase™ EPROM erasers and mid size UV viewing cabinets
Length (English) 5.31 in.
Length (Metric) 134.8 mm
Packaging Quantity 1 Ea.
Product Type UV Lamp Tube