Learn More
UVP Compact and Handheld UV Lamp Replacement Tubes
Product Code. 11786832
Change view
Click to view available options
Wavelength:
254 nm
365 nm
Wattage:
4 W
6 W
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength Wattage unitSize
11786832 365 nm 4 W Each
12812979 254 nm 4 W Each
11902041 254 nm 6 W Each
12862979 365 nm 6 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 11786832 Supplier UVP Supplier No. 34000501
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
For use with 4w and 6w UV lamps.
  • For use with 4w and 6w UV lamps
Specifications
Light Source UV-A
Voltage 230 V
Wattage 4 W
Wavelength 365 nm
Diameter (English) 0.62 in.
Diameter (Metric) 15.8 mm
For Use With (Equipment) EL-series UV lamp, 3UV™ lamp, UVGL-25, UVL-21, UVL-23R, UVL-23RW compact UV lamp and mid size UV viewing cabinets
Length (English) 6 in.
Length (Metric) 134.8 mm
Packaging Quantity 1 Ea.
Product Type UV Lamp Tube