Learn More
UVP UVL-56 Handheld UV Lamp
Click to view available options
Wavelength:
365 nm
Unit Size:
Each
Description
- Comfortable, ergonomically designed handle
- On/off switch is located on the back of the lamp
- Lamps are lightweight for easy handling
- Durable plastic housing
- Six watts
Specifications
Specifications
| Light Source | UV-A |
| Voltage | 230 V |
| Wavelength | 365 nm |
| Case Material | Plastic |
| Dimensions (L x W x H) | 378 x 81 x 64 mm |
| Model | UVL-56 Handheld UV Lamp |