Specifications
| Certifications/Compliance | EN 149, meets both the requirements for reusability (R) and the requirement for high dust exposure (D-marking, dolomite dust test), FFP3 protection level |
| Color | Gray |
| Filter Type | Activated carbon filter |
| Product Type | Particulate filtering form mask with exhalation valve |
| With Exhalation Valve | Yes |
| Quantity | 5/Pk. |
| Resistant To | Pollutants up to 30 times the limit and odors, gases and vapors below limit |