Specifications
| Certifications/Compliance | EN 149, Meets the requirements against high levels of dust (D-marking, dolomite dust test, Protection level FFP2 |
| Color | White |
| Filter Type | Classic cup style FFP2 |
| Product Type | Particulate filtering form mask with exhalation valve |
| With Exhalation Valve | Yes |
| Quantity | 15/Pk. |
| Resistant To | Pollutants up to 10 times the limit |
| With Faceseal | Yes |