Learn More
uvex C300 Foam Cut Protection Gloves
Shop All Uvex Products
Click to view available options
Size:
7
8
9
10
Unit Size:
Each
Description
- Gloves provide level 3/C blade cut resistance, abrasion resistance, and high tear resistance
- High-performance elastomer (HPE) coating on the palm and fingers for enhanced grip capabilities
- Soft-grip foam coating provides mechanical abrasion resistance
- Highly flexible with very good tactile sensation
- Uvex Climazone technology reduces excessive moisture and warmth
- 3D Ergo technology for a secure fit
- Silicone-free according to the imprint test
Specifications
Specifications
| Coating Material | HPE |
| Color | Gray |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Bamboo Viscose, HPPE, Polyamide, Glass |
| Product Type | Cut-resistant Gloves |
| Size | 9 |
| EN388 Standard | 3X42C |
| Glove Cuff Style | Knit |
| Series | C300 Foam |