Learn More
Uvex™ 1 Safety Shoe S2
Product Code. 15311213 Shop All Uvex Products
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
Each
Product Code. Size unitSize
15311213 39 Each
15381203 36 Each
15391203 37 Each
15301213 38 Each
15321213 40 Each
15331213 41 Each
15341213 42 Each
15351213 43 Each
15361213 44 Each
15371213 45 Each
15381213 46 Each
15391213 47 Each
15301223 48 Each
15311223 49 Each
14 options
Product Code. 15311213
Brand: Uvex™ 8544839
Only null left
Add to basket
Add to basket
Sporty, multifunctional safety shoe
  • Especially lightweight and flexible
  • 100% metal-free uvex xenova® toe cap of the latest generation, compact design, anatomical shape, good lateral stability, no thermal conductivity, non-magnetic
  • Made with a new last technology for extraordinary wearing comfort
  • Optimized comfort thanks to uvex climazone technology
  • Breathable, hydrophobic high-tech micro velor upper
  • Optimum ventilation thanks to Distance Mesh lining material
  • Suitable for allergy sufferers as made of synthetic materials
  • Individually adjustable, elastic lace with quick release
  • Almost seamless shaft construction to avoid pressure points
  • Removable comfort footbed, antistatic, with moisture transport system and additional heel and forefoot cushioning
  • Soft padded collar and padded flap
  • Special double-density polyurethane sole with excellent cushioning properties and very good slip resistance, free from substances that interfere with the wetting properties of the paint such as silicones, plasticisers, etc.
  • Meets the ESD requirements: Bleeder resistance <35 megohms
  • Certified with uvex medicare for individual orthopedic insoles and individual outsole modifications such as outsole heights, inner or outer heel elevations, roll-off aids and other solutions
  • Item No. 8544.8
  • Width 11
  • Standard EN ISO 20345: 2011 S2 SRC
  • TRUSTED_SUSTAINABILITY
    Specifications
    Certifications/Compliance ESD certified, EN ISO 20345: 2011 S2 SRC
    Color Black
    Design Loafer
    Material Microvelour
    Size 39
    Toe Material Composite
    Width (Metric) 11 mm
    Gender Unisex
    Ankle Style Trainers
    Waterproof No
    Product Type ESD Safety Shoe