Learn More
Jenway™ UV Ultra-Micro Cuvette
Product Code. 15401570
Change view
Click to view available options
Capacity (Metric):
100 μL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
15401570 100 μL Each
1 options
Product Code. 15401570
Brand: Jenway™ 8307088
Only null left
Add to basket
Add to basket
Quartz construction Jenway™ Ultra-Micro Cuvette is for use in both the UV and visible range.
  • For use with both UV and visible range
  • 10mm pathlength
  • Quartz construction
Specifications
Capacity (Metric) 100 μL
Material Quartz
Path Length 10 mm
For Use With (Equipment) Spectrophotometer
For Use With (Application) UV/Vis Spectrophotometry