Learn More
Cole-Parmer™ UV Tubes For Ultraviolet Equipment
Product Code. 11820390 Shop All Cole-Parmer Products
Change view
Click to view available options
Wavelength:
254 nm
312 nm
365 nm
Wattage:
4 W
6 W
15 W
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength Wattage Unit Size
11820390 254 nm 6 W Each
11800390 254 nm 4 W Each
11830390 365 nm 6 W Each
11880380 254 nm 15 W Each
11890380 312 nm 15 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11820390 Supplier Cole-Parmer™ Supplier No. 0981546
  • ONLINE EXCLUSIVE PRICE
    1400.00 NOK
    1290.83 NOK / Pack of 1
    Save 109.17 NOK
    7% Off
Estimated Shipment: 08-12-2026
Use these replacement UV tubes with ultraviolet lamps.
  • Replacement Ultraviolet Tubes
  • Product may contain trace amounts of mercury
  • Use caution with breakage, cleanup, shipping and/or disposal
Specifications
Light Source UV-C
Wattage 6 W
Wavelength 254 nm
Diameter (Metric) 15.5 mm
For Use With (Equipment) 6 W, 254 nm lamps
Length (Metric) 210.5 mm
Product Type UV Tube