Learn More
Cole-Parmer™ UV Tubes For Ultraviolet Equipment
Product Code. 11800390 Shop All Cole-Parmer Products
Change view
Click to view available options
Wavelength:
254 nm
312 nm
365 nm
Wattage:
4 W
6 W
15 W
Unit Size:
Each
Product Code. Wavelength Wattage unitSize
11800390 254 nm 4 W Each
11820390 254 nm 6 W Each
11830390 365 nm 6 W Each
11880380 254 nm 15 W Each
11890380 312 nm 15 W Each
5 options
Product Code. 11800390
Brand: Cole-Parmer™ 0981536
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these replacement UV tubes with ultraviolet lamps.
  • Replacement Ultraviolet Tubes
  • Product may contain trace amounts of mercury
  • Use caution with breakage, cleanup, shipping and/or disposal
Specifications
Light Source UV-C
Wattage 4 W
Wavelength 254 nm
Diameter (Metric) 15.5 mm
For Use With (Equipment) 4 W, 254 nm lamps
Length (Metric) 134.5 mm
Product Type UV Tube