Learn More
MP Biomedicals UV Tube, 8W 302 nm
Product Code. 15729220 Shop All MP Biomedicals Products
Change view
Click to view available options
Wavelength:
302 nm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength Unit Size
15729220 302 nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15729220 Supplier MP Biomedicals Supplier No. 11INSU9303
  • ONLINE EXCLUSIVE PRICE
    2265.00 NOK
    2155.02 NOK / Pack of 1
    Save 109.98 NOK
    4% Off
Estimated Shipment: 04-12-2026
Long-life UV tubes designed for MP Bio transilluminators; portable lamps or template tammer.
  • Long life.
  • Suitable for transilluminators, portable lamps, template tammer.
    • Compatible with compact transilluminators #INSU9300; INSU9301; INSU9302
  • Glass construction
  • For use in molecular biology
Specifications
Light Source UV
Wattage 8 W
Wavelength 302 nm
For Use With (Equipment) MP Bio Transilluminators, portable lamps, template tammer, compact transilluminators #INSU9300; INSU9301; INSU9302
Packaging Quantity 1 Ea.