Learn More
Syngene™ UV Safety Shield
Product Code. 16534351 Shop All Syngene Products
Change view
Click to view available options
Size:
Small
Large
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16534351 Small Each
16524351 Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16534351 Supplier Syngene™ Supplier No. UVSHIELDSMALL
  • ONLINE EXCLUSIVE PRICE
    2470.00 NOK
    2287.14 NOK / Pack of 1
    Save 182.86 NOK
    7% Off
Estimated Shipment: 26-01-2027
Safety shield designed to offer UV protection.
  • The wearing of safety equipment according to laboratory regulations is recommended
  • Small Shield Dimensions: 14 W x 19 in. H
Specifications
Product Line Syngene
For Use With (Application) UV Protection
Product Type Safety Shield
For Use With (Equipment) GBox F3 and GBox Chemi XRQ
Includes UV Safety Shield, 19 in. high
Size Small
Voltage 110/230 V