Learn More
Vilber Lourmat™ UV Lamp Stand
Product Code. p-8011084
Click to view available options
For Use With (Equipment):
15w UV lamp
4/8w Handheld UV Lamp
4/8w UV lamp
Unit Size:
Each
Product Code. 11840240
Brand: Vilber Lourmat™ 913020031
Rich and robust table stand for ultraviolet lamps 4/8 W.
  • Stand for UV lamps 4/8W
  • Double wavelength lamp
  • 254/365 nm Intensity 
  • Lamp 2 x 6 W
  • M, 2 table stand
Specifications
Type UV Lamp Stand
Description Stand for UV lamps 4/8W
For Use With (Equipment) 4/8w UV lamp