Learn More
Biochrom UV Grade Silica Semi-micro Cuvette with Stopper and Black Walls
10 mm Quartz cell with stopper and black walls. 800 μL working volume and 10 mm pathlength.
Brand:  Biochrom 80-2002-81
Additional Details : Weight : 0.02800kg
Product Code. 11390132
  • 7255.00 NOK / Each
Estimated Shipment: 26-07-2024
Description
  • Semi-micro cell with stopper and black walls
  • 10 mm pathlength
  • 4 mm internal width
  • 800 μL working volume
  • 12.5 mm x 48 mm external dimensions
  • Fits standard spectrophotometer cell holders
  • Used with visible/UV spectrophotometry
Specifications
Specifications
Type 5 Semi-Micro Cuvette
UV Grade Silica
1 Ea.
Dual Beam Ultrospec Spectrophotometers
800 μL
4 mm
10 mm
Spectrophotometric Measurements