Learn More
UT-B Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25962-1-AP-150UL
 View more versions of this product
Product Code. 16834495
  • 4836.10 NOK / 150µL
Description
The UT-B antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human UT-B. This antibody recognizes human, mouse antigen. The UT-B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
UT-B
Polyclonal
Unconjugated
150 μL
6563
IgG
Western Blot, ELISA
1187 μg/mL
Rabbit
Primary
Human, Mouse