Learn More
UT-B Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25962-1-AP-20UL
This item is not returnable.
View return policy
Description
The UT-B antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human UT-B. This antibody recognizes human, mouse antigen. The UT-B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| UT-B | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 6563 | |
| IgG |
| Western Blot, ELISA | |
| 1187 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |