missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP9X Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55054-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to USP9X. It has no cross reaction to USP9Y.
USP9X is an ubiquitin-specific protease involved in the removal and recycling of ubiquitin from proteins degraded in proteasomes. As a deubiquitinating enzyme, USP9x has a critical function in tight junction biogenesis by controlling the levels of the exchange factor for Arf6 (EFA6), a protein shown to facilitate tight junction formation during a narrow temporal window preceding the establishment of cell polarity. USP9X is also located on the X-chromosome but escapes X-inactivation, and defects in USP9X may cause gonadal degeneration in Turner syndrome.Specifications
| USP9X | |
| Polyclonal | |
| Unconjugated | |
| USP9X | |
| Deubiquitinating enzyme FAF X, DFFRX, FAF, FAM, Fat facets in mammals, hFAM, Ubiquitin thiolesterase FAF X, USP9, USP9X | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22284, 363445, 8239 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| 0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P70398, Q93008 | |
| Usp9x | |
| Peptide | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction