Learn More
USP8 Mouse anti-Human, Unconjugated, Clone: 1E12E3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67321-1-IG-20UL
 View more versions of this product
Product Code. 16849885
  • 1710.33 NOK / 20µL
Description
The USP8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human USP8. This antibody recognizes human, mouse antigen. The USP8 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
USP8
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1E12E3
Unconjugated
20 μL
9101
IgG1