Learn More
USP8 Mouse anti-Human, Unconjugated, Clone: 1E12E3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67321-1-IG-150UL
This item is not returnable.
View return policy
Description
The USP8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human USP8. This antibody recognizes human, mouse antigen. The USP8 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| USP8 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1E12E3 | |
| Unconjugated | |
| 150 μL | |
| 9101 | |
| IgG1 |