Learn More
USP44 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15521-1-AP-20UL
This item is not returnable.
View return policy
Description
Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP44 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes.[supplied by OMIM, Mar 2008].Specifications
| USP44 | |
| Polyclonal | |
| Unconjugated | |
| USP44 | |
| Deubiquitinating enzyme 44, Ubiquitin thiolesterase 44, USP44 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 327799, 84101 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8C2S0, Q9H0E7 | |
| Usp44 | |
| USP44 Fusion Protein Ag7888 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |