Description
The USP25 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human USP25. This antibody recognizes human, mouse, rat antigen. The USP25 antibody has been validated for the following applications: WB, IHC, ELISA analysis.
Specifications
Specifications
| Antigen | USP25 |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Clone | 2B6A2 |
| Concentration | 1000 μg/mL |
| Conjugate | Unconjugated |
| Gene Symbols | USP21 |
| Host Species | Mouse |
| Immunogen | Recombinant Protein |
| Purification Method | Protein A purification |
| Show More |