missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ US12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144429
This item is not returnable.
View return policy
Description
US12 Polyclonal Antibody for Western Blot, ELISA
Plays a role in the inhibition of host immune response. Binds specifically to transporters associated with antigen processing (TAP), thereby blocking peptide-binding and translocation by TAP as well as subsequent loading of peptides onto MHC class I molecules. Empty MHC I molecules are retained in the endoplasmic reticulum and ultimately directed to proteasomal degradation. In consequence, infected cells are masked for immune recognition by cytotoxic T-lymphocytes.
Specifications
| US12 | |
| Polyclonal | |
| Unconjugated | |
| US12 | |
| ICP47 protein; US12 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 2703441 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 1.249 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P03170 | |
| US12 | |
| Recombinant Human herpesvirus 1 ICP47 protein (US12) (1-88aa). | |
| 100 μL | |
| Primary | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction