Learn More
MedChemExpress Uridine triphosphate
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Molecular Weight (g/mol) | 484.14 g/mol |
| Product Type | Natural Products |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Purity | 98.0% |
| Quantity | 50 mg |
| Solubility Information | H2O : ≥ 150 mg/mL (309.83 mM) |
| For Use With (Application) | Cancer-Kinase/protease |
| CAS | 63-39-8 |
| Synonym | UTP∣ Uridine 5'-triphosphate |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.