Learn More
Urea, Multi-Compendial, U.S.P., J.T. Baker™
Product Code. 15528274
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
Product Code. 15528274
Brand: Avantor J.T.Baker 420301
Specifications
CAS 57-13-6
Packaging Poly Bottle
Molecular Formula CH4N2O
Quantity 500 g
Specific Gravity 1L = 1.32kg
Synonym carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILES C(=O)(N)N
IUPAC Name urea
Molecular Weight (g/mol) 60.056
PubChem CID 1176
ChEBI CHEBI:48376
Formula Weight 60.06
Grade Multi-Compendial/USP
Chemical Name or Material Urea