Learn More
URB1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Product Code. 16859504 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16859504 20 μL 20µL
16849504 150 μL 150µL
2 options
Product Code. 16859504
Brand: Proteintech 200231AP20UL
Rabbit Polyclonal Antibody
The antibody is specific to URB1.
URB1 gene ontology annotations related to this gene include maturation of 5.8S rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA); maturation of LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA).
Specifications
Antigen URB1
Applications Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene URB1
Gene Accession No. O60287, Q571H0
Gene Alias C21orf108, KIAA0539, URB1
Gene Symbols URB1
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 207932, 9875
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG