Description
With injector unit vials, test tubes, autosampler tubes clearance 190mm 531mm (w) x 475mm (d) x 260mm (h) Miele
Specifications
Specifications
| For Use With (Equipment) | Narrow-Necked Glassware |
| Quantity | 1 Each |
| Dimensions (L x W x H) | 475 x 531 x 170 mm |
| Depth (Metric) | 475 mm |
| Includes | 34 Nozzles (E351) 4 x 160mm, 34 clips for nozzle 4 x 160mm (E 353) |
| Product Type | Upper Basket |
| Height (Metric) | 170 mm |
| Width (Metric) | 531 mm |