missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UPK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21149-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes one of the proteins of the highly conserved urothelium-specific integral membrane proteins of the asymmetric unit membrane which forms urothelium apical plaques in mammals. The asymmetric unit membrane is believed to strengthen the urothelium by preventing cell rupture during bladder distention. The encoded protein is expressed in the peripheral blood of bladder cancer patients with transitional cell carcinomas.Specifications
UPK2 | |
Polyclonal | |
Unconjugated | |
UPK2 | |
UP2, UPII, upk 2, UPK2, uroplakin 2, Uroplakin II | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22269, 689093, 7379 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00526, P38575 | |
UPK2 | |
UPK2 Fusion Protein Ag15694 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |