Learn More
Kinesis™ PEEK Chromatography Tubing
Description
An excellent replacement for stainless steel tubing
Can be used in place of stainless steel tubing in most liquid analytical systems
- Biocompatible, inert and easy to cut
- Great for high-pressure applications
- Wide choice of outer and inner diameters
- Many sizes color-coded for easier inner diameter identification
Specifications
Specifications
| Type | Tubing |
| Color | Blue |
| For Use With (Application) | High-Pressure Applications |
| Diameter (English) Inner | 0.01 in. |
| Overall Length (English) | 50 ft. |
| Material | PEEK |
| Maximum Pressure | 7000 psi (483 bar) |