Learn More
Kinesis™ PEEK Chromatography Tubing
Product Code. 13009902
Change view
Click to view available options
Diameter (English) Inner:
0.005 in.
0.007 in.
0.01 in.
0.02 in.
0.03 in.
0.04 in.
Overall Length (English):
5 ft.
50 ft.
100 ft.
Maximum Pressure:
5000 psi (345 bar)
7000 psi
7000 psi (483 bar)
Color:
Blue
Green
Natural
Orange
Red
Yellow
Unit Size:
Each
Product Code. Diameter (English) Inner Overall Length (English) Maximum Pressure Color unitSize
13009902 0.007 in. 50 ft. 7000 psi (483 bar) Yellow Each
13069892 0.005 in. 5 ft. 7000 psi (483 bar) Red Each
13039882 0.01 in. 5 ft. 7000 psi (483 bar) Natural Each
13099882 0.02 in. 5 ft. 7000 psi (483 bar) Orange Each
13029892 0.03 in. 5 ft. 7000 psi (483 bar) Green Each
13039902 0.04 in. 5 ft. 5000 psi (345 bar) Natural Each
13059882 0.01 in. 50 ft. 7000 psi (483 bar) Blue Each
13049892 0.03 in. 100 ft. 7000 psi Green Each
8 options
Product Code. 13009902
Brand: Kinesis™ 0200633
Only null left
Add to basket
Add to basket
An excellent replacement for stainless steel tubing
Can be used in place of stainless steel tubing in most liquid analytical systems
  • Biocompatible, inert and easy to cut
  • Great for high-pressure applications
  • Wide choice of outer and inner diameters
  • Many sizes color-coded for easier inner diameter identification
Specifications
Type Tubing
Color Yellow
For Use With (Application) High-Pressure Applications
Diameter (English) Inner 0.007 in.
Overall Length (English) 50 ft.
Material PEEK
Maximum Pressure 7000 psi (483 bar)