Learn More
Kinesis™ Peek Low-Pressure Union Assembly
Description
Make solid connections in low-pressure applications
- Manufactured from inert polymers
- 1/4-28 flat-bottom ports
- Max pressure of 1000 psi (69 bar)
- 0.030 in. thru-hole diameter
- 0.41 μL swept volume
Specifications
Specifications
| Type | Union |
| Connections | 2F-125/1P-553, 6-32 |
| Material | PEEK |
| Color | Natural |
| For Use With (Equipment) | MicroTight Sleeves |