Learn More
Kinesis™ LiteTouch™ Ferrule System; 1/8 in. PEEK Nut
Click to view available options
For Use With (Equipment):
LiteTouch Ferrule System
Unit Size:
Pack of 10
Specifications
Specifications
| Type | Nut |
| Description | Fits 1/8 in. O.D. tubing |
| Thread Size | 10 to 32 UNF |
| For Use With (Equipment) | LiteTouch Ferrule System |
| Material | PEEK |