Learn More
Kinesis™ Flangeless Fittings: Ferrules, Tefzel
Product Code. 12987589
Change view
Click to view available options
Color:
Natural
Yellow
For Use With (Equipment):
1/16 in. OD Tubing
1/16 in. OD Tubing, Male Nuts
1/8 in. OD Tubing, Male Nuts
Length (English):
0.22 in.
Not Applicable
Width (English):
0.18 in.
Not Applicable
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color For Use With (Equipment) Length (English) Width (English) Unit Size
12987589 Yellow 1/16 in. OD Tubing Not Applicable Not Applicable Pack of 10
12917579 Natural 1/16 in. OD Tubing, Male Nuts 0.22 in. 0.18 in. Pack of 10
12987609 Natural 1/8 in. OD Tubing, Male Nuts Not Applicable Not Applicable Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12987589 Supplier Kinesis™ Supplier No. 0201374
  • ONLINE EXCLUSIVE PRICE
    746.00 NOK
    689.66 NOK / Pack of 10
    Save 56.34 NOK
    7% Off
Estimated Shipment: 10-12-2026
No need to flange tubing
Excellent for connecting rigid or semi-rigid tubing
Specifications
Type Flangeless Ferrule
Length (English) Not Applicable
Width (English) Not Applicable
Material ETFE
Color Yellow
For Use With (Equipment) 1/16 in. OD Tubing