Learn More
Novus Biologicals™ UNveil Unstained Protein Ladder
Description
The Unveil Unstained Protein Ladder is a mixture of 12 unstained recombinant proteins covering a wide range of molecular weights from 10 to 200 kDa. The proteins resolve into clearly identifiable bands when separated on the SDS-PAGE gel (MES buffer), with the intensified 25kDa and 85kDa protein bands serving as the two reference bands. The Unveil Unstained Protein Ladder is designed for monitoring protein separation during SDS-polyacrylamide gel electrophoresis and verifying Western transfer efficiency on membranes (PVDF, nylon, or nitrocellulose) and for approximating the size of proteins. The ladder is supplied in the gel loading buffer and is ready to use.
Specifications
Specifications
| Formulation | 20 mM Tris-phosphate (pH 7.5), 2 % SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, 15 % (v/v) Glycerol. |
| For Use With (Application) | 3 μL or 5 μL per loading for clear visualization during electrophoresis on 15-well or 10-well mini-gel, respectively. 2.5 μL per well for general Western transferring |
| Quantity | 500 μL |