Learn More
Buerkle™ Uno Sampler
Product Code. 10228420
Change view
Click to view available options
Length (Metric):
55 cm
85 cm
150 cm
Unit Size:
Each
Product Code. Length (Metric) unitSize
10228420 85 cm Each
10616721 55 cm Each
10780841 150 cm Each
3 options
Product Code. 10228420
Brand: Buerkle™ 53161085
Only null left
Add to basket
Add to basket
Buerkle™ Uno Sampler comes with one closed sample chamber for point sampling from a predetermined sampling depth.
  • Outer tube stainless steel AISI 316L (1.4404), sample chambers made of PTFE
  • With colour coding system
  • Smallest possible gap between the inner and the outer tube
  • Easy cleaning, removable tip
  • Colour coding (Multi and Uno sampler)
  • Ultra-pure materials
  • Excellent processing, robust model
TRUSTED_SUSTAINABILITY
Specifications
Product Type Sampler
Material Stainless Steel (V4A), PTFE
Autoclavable Autoclavable
Includes 1 Collection Chamber
Handle Type T-shape
Capacity (Metric) 17 mL
Certifications/Compliance DIN
For Use With (Application) Ideal for Coarse Grains and Larger Quantities, Foodstuff
Length (Metric) 85 cm