Learn More
Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers
Product Code. 10474992 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Flask Clamp
Flask Clamp Kit
Microplate Clamp
Holds:
1 L Erlenmeyer Flask
125 mL Erlenmeyer Flask
2 L Erlenmeyer Flask
25 mL Erlenmeyer Flask
250/300 mL Erlenmeyer Flask
2800 mL Fernbach Flask
4 L Erlenmeyer Flask
5 L Erlenmeyer Flask
50 mL Erlenmeyer Flask
500 mL Erlenmeyer Flask
6 L Erlenmeyer Flask
Flasks
Microplates
Unit Size:
Each
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Holds Unit Size
10474992 Flask Clamp 25 mL Erlenmeyer Flask Each
10525433 Flask Clamp 50 mL Erlenmeyer Flask Each
10729273 Flask Clamp 125 mL Erlenmeyer Flask Each
11821271 Flask Clamp 250/300 mL Erlenmeyer Flask Each
10535433 Flask Clamp 500 mL Erlenmeyer Flask Each
10084032 Flask Clamp 1 L Erlenmeyer Flask Each
10452902 Flask Clamp 2 L Erlenmeyer Flask Each
10462902 Flask Clamp 2800 mL Fernbach Flask Each
11826763 Microplate Clamp Microplates Each
12682785 Flask Clamp 4 L Erlenmeyer Flask Each
12692785 Flask Clamp 6 L Erlenmeyer Flask Each
11910398 Flask Clamp 5 L Erlenmeyer Flask Each
10338922 Flask Clamp Kit Flasks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 10474992 Supplier Thermo Scientific™ Supplier No. 236010
  • ONLINE EXCLUSIVE PRICE
    730.00 NOK
    682.04 NOK / Pack of 1
    Save 47.96 NOK
    6% Off
In Stock
Add to basket
Edge
Add to basket
Secure vessels using these universal platform clamps for orbital shakers.
Secure vessels using Thermo Scientific™ Universal Platform Clamps for MaxQ™ High Performance Orbital Shakers. With exceptional innovation, reliability and accuracy, these orbital shakers, platforms and accessories are the optimal choice for a busy lab.
Flask Clamp Starter Kit includes:
  • (1) microplate clamp
  • (2) 25mL flask clamps
  • (2) 50mL flask clamps
  • (2) 125mL flask clamps
  • (4) 250/300mL flask clamps
  • (4) 500mL flask clamps
  • (2) 1L flask clamps
  • (2) 2L flask clamps
  • Replacement screws
Note:
Replacement screws also available separately.
TRUSTED_SUSTAINABILITY
Specifications
Holds 25 mL Erlenmeyer Flask
Product Type Flask Clamp
For Use With (Equipment) MaxQ™ 8000 Shaker